siRNA Details
| Gene Symbol: BRD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317951.1 | NP_001304880.1 | 4 | 686 | ||
| NR_134296.1 | 4 | 903 | ||||
| GenBank mRNA | AK023503.1 | - | 4 | 664 | ||
| AK024392.1 | - | 1 | 402 | |||
| AK301987.1 | - | 4 | 518 | |||
| BC007217.1 | - | 3 | 410 | |||
| BC041590.1 | - | 2 | 253 | 5428011 | ||
| BX648703.1 | - | 4 | 905 | |||

BRD9
Genomic Map