siRNA Details
Gene Symbol: LINC00116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027063.1 | 2 | 271 | |||
GenBank mRNA | AF504645.1 | - | 2 | 271 | ||
BC064430.1 | - | 3 | 703 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027063.1 | 2 | 271 | |||
GenBank mRNA | AF504645.1 | - | 2 | 271 | ||
BC064430.1 | - | 3 | 703 |