siRNA Details
| Gene Symbol: CENPBD1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026052.1 | 2 | 2689 | |||
| GenBank mRNA | AK098293.1 | - | 1 | 1694 | ||
| BC043401.1 | - | 2 | 2628 | |||
| BC098570.1 | - | 1 | 2199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CENPBD1P1| Gene Symbol: CENPBD1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026052.1 | 2 | 2689 | |||
| GenBank mRNA | AK098293.1 | - | 1 | 1694 | ||
| BC043401.1 | - | 2 | 2628 | |||
| BC098570.1 | - | 1 | 2199 | |||