siRNA Details
| Gene Symbol: CENPBD1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026052.1 | 2 | 3239 | |||
| GenBank mRNA | AA418278.1 | - | 1 | 314 | ||
| BC098570.1 | - | 1 | 2749 | |||
| Gene Symbol: LOC101929188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134628.1 | 2 | 1349 | |||
| GenBank mRNA | AF148948.2 | - | 1 | 379 | ||
| AF148949.2 | - | 1 | 379 | |||
| BC044947.1 | - | 2 | 1240 | |||
| Gene Symbol: LOC105378948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF148950.2 | - | 1 | 546 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF244571.1 | - | 1 | 287 | Chr. 3 - 184954501 - 184955984 [+] on Build GRCh38 | |

CENPBD1P1
Genomic Map