siRNA Details
| Gene Symbol: HIGD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002780.1 | 3 | 975 | |||
| GenBank mRNA | BC040890.1 | - | 3 | 975 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIGD2B| Gene Symbol: HIGD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002780.1 | 3 | 975 | |||
| GenBank mRNA | BC040890.1 | - | 3 | 975 | ||