siRNA Details
| Gene Symbol: HIGD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002780.1 | 1, 2 | 341 | |||
| GenBank mRNA | BC040890.1 | - | 1, 2 | 341 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM564128.1 | - | 1, 2 | 361 | Chr. 15 - 72675889 - 72686150 [-] on Build GRCh38 | |

HIGD2B
Genomic Map