siRNA Details
| Gene Symbol: AOC4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002773.1 | 1 | 1780 | |||
| GenBank mRNA | AF047485.1 | - | 1 | 1780 | ||
| AF047486.1 | - | 2 | 569 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AOC4P| Gene Symbol: AOC4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002773.1 | 1 | 1780 | |||
| GenBank mRNA | AF047485.1 | - | 1 | 1780 | ||
| AF047486.1 | - | 2 | 569 | |||