siRNA Details
| Gene Symbol: JMJD7-PLA2G4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB209150.1 | - | 7 | 3418 | ||
| BC128426.1 | - | 1 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

JMJD7-PLA2G4B| Gene Symbol: JMJD7-PLA2G4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB209150.1 | - | 7 | 3418 | ||
| BC128426.1 | - | 1 | 274 | |||