siRNA Details
| Gene Symbol: LOC606724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002454.2 | 1 | 405 | |||
| GenBank mRNA | U35342.1 | - | 1 | 405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC606724| Gene Symbol: LOC606724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002454.2 | 1 | 405 | |||
| GenBank mRNA | U35342.1 | - | 1 | 405 | ||