siRNA Details
| Gene Symbol: LOC606724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002454.2 | 1 | 72 | |||
| GenBank mRNA | U35342.1 | - | 1 | 72 | ||
| Gene Symbol: CORO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017022885.1 | XP_016878374.1 | 10 | 1765 | ||
| XM_017022886.1 | XP_016878375.1 | 10 | 1587 | |||
| GenBank mRNA | AK097964.1 | - | 2 | 1916 | ||
| AK123401.1 | - | 3 | 2571 | |||

LOC606724
Genomic Map