siRNA Details
| Gene Symbol: FLJ37453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024279.1 | 2 | 1125 | |||
| GenBank mRNA | AK094772.1 | - | 1 | 39 | ||
| AK124018.1 | - | 2 | 1126 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ37453| Gene Symbol: FLJ37453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024279.1 | 2 | 1125 | |||
| GenBank mRNA | AK094772.1 | - | 1 | 39 | ||
| AK124018.1 | - | 2 | 1126 | |||