siRNA Details
| Gene Symbol: CST13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001279.2 | 4 | 1118 | |||
| GenBank mRNA | BC024006.1 | - | 4 | 1123 | ||
| BC028368.1 | - | 4 | 1123 | |||
| BC033934.1 | - | 4 | 1113 | |||
| BC041171.1 | - | 4 | 1097 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB522378.1 | - | 3 | 434 | Chr. 20 - 23521092 - 23542012 [+] on Build GRCh38 | |

CST13P
Genomic Map