siRNA Details
| Gene Symbol: CST13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001279.2 | 3 | 778 | |||
| GenBank mRNA | BC024006.1 | - | 3 | 783 | ||
| BC028368.1 | - | 3 | 783 | |||
| BC033934.1 | - | 3 | 773 | |||
| BC041171.1 | - | 3 | 757 | |||
| FJ895881.1 | - | 2 | 178 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB522378.1 | - | 2 | 94 | Chr. 20 - 23521092 - 23542012 [+] on Build GRCh38 | |

CST13P
Genomic Map