siRNA Details
| Gene Symbol: ADAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002224.2 | 2 | 1801 | |||
| GenBank mRNA | AK093400.1 | - | 2 | 1801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADAM6| Gene Symbol: ADAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002224.2 | 2 | 1801 | |||
| GenBank mRNA | AK093400.1 | - | 2 | 1801 | ||