siRNA Details
| Gene Symbol: GOLGA2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120609.1 | 2 | 886 | |||
| GenBank mRNA | AK097852.1 | - | 3 | 383 | ||
| AK125739.1 | - | 2 | 886 | |||
| AK309583.1 | - | 2 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GOLGA2P6| Gene Symbol: GOLGA2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120609.1 | 2 | 886 | |||
| GenBank mRNA | AK097852.1 | - | 3 | 383 | ||
| AK125739.1 | - | 2 | 886 | |||
| AK309583.1 | - | 2 | 479 | |||