siRNA Details
| Gene Symbol: CLLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025233.2 | NP_001020404.1 | 2 | 3910 | ||
| NR_027932.1 | 3 | 2796 | ||||
| NR_027933.1 | 2 | 2239 | ||||
| GenBank mRNA | AJ845162.1 | - | 2 | 2239 | ||
| AJ845163.1 | - | 3 | 2796 | |||

CLLU1
Genomic Map