siRNA Details
| Gene Symbol: CLLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027932.1 | 1 | 136 | |||
| NR_027933.1 | 1 | 136 | ||||
| GenBank mRNA | AJ845162.1 | - | 1 | 136 | ||
| AJ845163.1 | - | 1 | 136 | |||
| AJ845164.1 | - | 1 | 136 | |||
| AJ845167.1 | - | 1 | 136 | |||

CLLU1
Genomic Map