siRNA Details
| Gene Symbol: SERTAD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024337.1 | 2 | 557 | |||
| GenBank mRNA | BC066352.1 | - | 2 | 556 | ||
| BC094794.1 | - | 1 | 2457 | |||
| BP422084.1 | - | 2 | 469 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERTAD4-AS1| Gene Symbol: SERTAD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024337.1 | 2 | 557 | |||
| GenBank mRNA | BC066352.1 | - | 2 | 556 | ||
| BC094794.1 | - | 1 | 2457 | |||
| BP422084.1 | - | 2 | 469 | |||