siRNA Details
| Gene Symbol: SERTAD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024337.1 | 1 | 172 | |||
| GenBank mRNA | BC066352.1 | - | 1 | 171 | ||
| BC094794.1 | - | 1 | 208 | |||
| BP422084.1 | - | 1 | 84 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG709605.1 | - | 1 | 174 | Chr. 1 - 210231479 - 210234124 [-] on Build GRCh38 | |

SERTAD4-AS1
Genomic Map