siRNA Details
| Gene Symbol: MIR9-3HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015411.1 | 7 | 2655 | |||
| NR_133001.1 | 7 | 3235 | ||||
| NR_133002.1 | 6 | 2536 | ||||
| NR_133003.1 | 7 | 2570 | ||||
| GenBank mRNA | AK054710.1 | - | 1 | 272 | ||
| AK096359.1 | - | 1 | 771 | |||

MIR9-3HG
Genomic Map