siRNA Details
| Gene Symbol: CSN1S2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003720.1 | 5, 6 | 196 | |||
| GenBank mRNA | AY154892.1 | - | 5, 6 | 196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSN1S2AP| Gene Symbol: CSN1S2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003720.1 | 5, 6 | 196 | |||
| GenBank mRNA | AY154892.1 | - | 5, 6 | 196 | ||