siRNA Details
| Gene Symbol: CSN1S2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003720.1 | 13 | 555 | |||
| GenBank mRNA | AY154892.1 | - | 13 | 555 | ||
| BC128170.1 | - | 10 | 429 | |||
| BC128173.1 | - | 10 | 429 | |||
| BC141914.1 | - | 10 | 429 | |||
| BC150502.1 | - | 10 | 429 | |||

CSN1S2AP
Genomic Map