siRNA Details
| Gene Symbol: HCG26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002812.3 | 1 | 1036 | |||
| GenBank mRNA | L29376.1 | - | 1 | 1035 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCG26| Gene Symbol: HCG26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002812.3 | 1 | 1036 | |||
| GenBank mRNA | L29376.1 | - | 1 | 1035 | ||