siRNA Details
| Gene Symbol: LINC00662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027301.1 | 3 | 2020 | |||
| GenBank mRNA | AK055559.1 | - | 3 | 1750 | ||
| BC045720.1 | - | 3 | 1779 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00662| Gene Symbol: LINC00662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027301.1 | 3 | 2020 | |||
| GenBank mRNA | AK055559.1 | - | 3 | 1750 | ||
| BC045720.1 | - | 3 | 1779 | |||