siRNA Details
| Gene Symbol: LINC00887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024480.1 | 3 | 2095 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00887| Gene Symbol: LINC00887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024480.1 | 3 | 2095 | |||