siRNA Details
| Gene Symbol: TMPRSS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026734.1 | 5 | 626 | |||
| GenBank mRNA | AK131211.1 | - | 5 | 626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS7| Gene Symbol: TMPRSS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026734.1 | 5 | 626 | |||
| GenBank mRNA | AK131211.1 | - | 5 | 626 | ||