siRNA Details
| Gene Symbol: SDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079653.1 | NP_001073121.1 | 1 | 76 | ||
| GenBank mRNA | AL833354.1 | - | 1 | 73 | ||
| DB065853.1 | - | 1 | 72 | |||
| DB454012.1 | - | 1 | 76 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDK1| Gene Symbol: SDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079653.1 | NP_001073121.1 | 1 | 76 | ||
| GenBank mRNA | AL833354.1 | - | 1 | 73 | ||
| DB065853.1 | - | 1 | 72 | |||
| DB454012.1 | - | 1 | 76 | |||