siRNA Details
Gene Symbol: SH3RF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029193.1 | 1 | 2559 | |||
GenBank mRNA | AF035291.1 | - | 1 | 1369 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SH3RF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029193.1 | 1 | 2559 | |||
GenBank mRNA | AF035291.1 | - | 1 | 1369 |