siRNA Details
| Gene Symbol: SH3RF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029193.1 | 1 | 2606 | |||
| GenBank mRNA | AF035291.1 | - | 1 | 1416 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH3RF3-AS1| Gene Symbol: SH3RF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029193.1 | 1 | 2606 | |||
| GenBank mRNA | AF035291.1 | - | 1 | 1416 | ||