siRNA Details
| Gene Symbol: SH3RF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029193.1 | 1 | 2028 | |||
| GenBank mRNA | AF035291.1 | - | 1 | 838 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX099992.1 | - | 1 | 206 | Chr. 2 - 109127616 - 109128297 [-] on Build GRCh38 | |

SH3RF3-AS1
Genomic Map