siRNA Details
| Gene Symbol: LINC00905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024335.2 | 2 | 197 | |||
| GenBank mRNA | AK058057.1 | - | 2 | 156 | ||
| BC069223.1 | - | 2 | 129 | |||
| BC104221.2 | - | 1 | 94 | |||
| DB461577.1 | - | 2 | 197 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000588117.6 | - | 2 | 256 | Chr. 19 - 16033654 - 16042116 [+] on Build GRCh38 | |

LINC00905
Genomic Map