siRNA Details
| Gene Symbol: LINC00905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024335.2 | 2 | 301 | |||
| GenBank mRNA | AK058057.1 | - | 2 | 260 | ||
| BC069223.1 | - | 2 | 233 | |||
| BC104221.2 | - | 1 | 198 | |||
| DB461577.1 | - | 2 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00905| Gene Symbol: LINC00905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024335.2 | 2 | 301 | |||
| GenBank mRNA | AK058057.1 | - | 2 | 260 | ||
| BC069223.1 | - | 2 | 233 | |||
| BC104221.2 | - | 1 | 198 | |||
| DB461577.1 | - | 2 | 301 | |||