siRNA Details
| Gene Symbol: LINC00905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024335.2 | 3 | 757 | |||
| NR_110321.1 | 3 | 467 | ||||
| GenBank mRNA | AK058057.1 | - | 3 | 716 | ||
| BC031284.1 | - | 3 | 467 | |||
| BC069223.1 | - | 3 | 689 | |||
| BC104221.2 | - | 2 | 654 | |||

LINC00905
Genomic Map