siRNA Details
| Gene Symbol: LINC00905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024335.2 | 2 | 534 | |||
| NR_110321.1 | 2 | 244 | ||||
| GenBank mRNA | AK058057.1 | - | 2 | 493 | ||
| BC031284.1 | - | 2 | 244 | |||
| BC069223.1 | - | 2 | 466 | |||
| BC104221.2 | - | 1 | 431 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI562021.1 | - | 2 | 249 | Chr. 19 - 16034120 - 16041522 [+] on Build GRCh38 | |

LINC00905
Genomic Map