siRNA Details
Gene Symbol: LINC00426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024464.2 | 5 | 1535 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024464.2 | 5 | 1535 |