siRNA Details
| Gene Symbol: LINC00426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024464.2 | 5 | 1491 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00426| Gene Symbol: LINC00426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024464.2 | 5 | 1491 | |||