siRNA Details
Gene Symbol: LOC100101148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104007.1 | 4 | 592 | |||
GenBank mRNA | BC070189.1 | - | 4 | 592 |
Gene Symbol: LOC541473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003602.3 | 5 | 730 | |||
GenBank mRNA | BC022013.2 | - | 5 | 684 | ||
BC067783.1 | - | 5 | 703 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG722503.1 | - | 5 | 690 | Chr. 7 - 75392097 - 75395415 [-] on Build GRCh38 |
