siRNA Details
| Gene Symbol: LINC00930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021493.1 | 3 | 1236 | |||
| GenBank mRNA | AK093148.1 | - | 1081 | |||
| AL045331.1 | - | 3 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00930| Gene Symbol: LINC00930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021493.1 | 3 | 1236 | |||
| GenBank mRNA | AK093148.1 | - | 1081 | |||
| AL045331.1 | - | 3 | 436 | |||