siRNA Details
| Gene Symbol: LINC00930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021493.1 | 1 | 79 | |||
| GenBank mRNA | DB045163.1 | - | 1 | 227 | ||
| DB061727.1 | - | 1 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00930| Gene Symbol: LINC00930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021493.1 | 1 | 79 | |||
| GenBank mRNA | DB045163.1 | - | 1 | 227 | ||
| DB061727.1 | - | 1 | 140 | |||