siRNA Details
Gene Symbol: LINC00930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021493.1 | 1 | 141 | |||
GenBank mRNA | DB045163.1 | - | 1 | 289 | ||
DB061727.1 | - | 1 | 202 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021493.1 | 1 | 141 | |||
GenBank mRNA | DB045163.1 | - | 1 | 289 | ||
DB061727.1 | - | 1 | 202 |