siRNA Details
Gene Symbol: LINC00930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021493.1 | 1 | 603 | |||
GenBank mRNA | AK093148.1 | - | 382 | |||
AL045331.1 | - | 1 | 40 | |||
DQ595207.1 | - | 11 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021493.1 | 1 | 603 | |||
GenBank mRNA | AK093148.1 | - | 382 | |||
AL045331.1 | - | 1 | 40 | |||
DQ595207.1 | - | 11 |