siRNA Details
| Gene Symbol: COL6A4P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027927.1 | 3 | 1428 | |||
| GenBank mRNA | AB299979.1 | - | 3 | 1428 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DW009796.1 | - | 1 | 284 | Chr. 3 - 15173927 - 15175059 [-] on Build GRCh38 | |

COL6A4P1
Genomic Map