siRNA Details
| Gene Symbol: DKFZp434L192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026929.1 | 1 | 916 | |||
| GenBank mRNA | AL713776.1 | - | 1 | 915 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 DKFZp434L192
DKFZp434L192| Gene Symbol: DKFZp434L192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026929.1 | 1 | 916 | |||
| GenBank mRNA | AL713776.1 | - | 1 | 915 | ||
 Back To Top
 Back To Top