siRNA Details
| Gene Symbol: LOC148413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015434.1 | 3 | 1590 | |||
| GenBank mRNA | AK024040.1 | - | 3 | 1564 | ||
| AK092572.1 | - | 1 | 2083 | |||
| BC068600.1 | - | 3 | 1547 | |||
| BC080546.1 | - | 3 | 1546 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC148413| Gene Symbol: LOC148413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015434.1 | 3 | 1590 | |||
| GenBank mRNA | AK024040.1 | - | 3 | 1564 | ||
| AK092572.1 | - | 1 | 2083 | |||
| BC068600.1 | - | 3 | 1547 | |||
| BC080546.1 | - | 3 | 1546 | |||