siRNA Details
| Gene Symbol: RRN3P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003370.2 | 1, 2 | 61 | |||
| GenBank mRNA | BX101439.1 | - | 1, 2 | 151 | ||
| CN397262.1 | - | 1, 2 | 68 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RRN3P1| Gene Symbol: RRN3P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003370.2 | 1, 2 | 61 | |||
| GenBank mRNA | BX101439.1 | - | 1, 2 | 151 | ||
| CN397262.1 | - | 1, 2 | 68 | |||