siRNA Details
| Gene Symbol: MIR202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030170.1 | 1 | 24 | |||
| Gene Symbol: MIR202HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108078.1 | 2 | 200 | |||

MIR202
Genomic Map