siRNA Details
| Gene Symbol: MIR202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030170.1 | 1 | 65 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830125.1 | - | 1 | 317 | Chr. 10 - 133247148 - 133247872 [-] on Build GRCh38 | |

MIR202
Genomic Map