siRNA Details
| Gene Symbol: LOC730101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024403.1 | 2 | 3602 | |||
| NR_024405.1 | 3 | 3372 | ||||
| GenBank mRNA | AK091117.1 | - | 3 | 3369 | ||
| AK095359.1 | - | 2 | 2655 | |||
| AK095807.1 | - | 3 | 2445 | |||
| AL049354.1 | - | 1 | 1073 | |||

LOC730101
Genomic Map