siRNA Details
| Gene Symbol: TMEM67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024522.1 | 29 | 3264 | |||
| GenBank mRNA | BC032835.2 | - | 29 | 3182 | 4825770 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM67| Gene Symbol: TMEM67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024522.1 | 29 | 3264 | |||
| GenBank mRNA | BC032835.2 | - | 29 | 3182 | 4825770 | |