siRNA Details
| Gene Symbol: KIAA1324L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291990.1 | NP_001278919.1 | 1, 2 | 134 | ||
| GenBank mRNA | AK303619.1 | - | 1, 2 | 136 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA1324L| Gene Symbol: KIAA1324L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291990.1 | NP_001278919.1 | 1, 2 | 134 | ||
| GenBank mRNA | AK303619.1 | - | 1, 2 | 136 | ||